Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761974.1 | XP_008760196.1 | 4 | 2856 | ||
GenBank mRNA | FQ209892.1 | - | 1 | 28 |