Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIA2 SIRNA Assays ›
Gene Symbol:
PPFIA2
Gene Name:
PTPRF interacting protein alpha 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 81257975 - 81759748 on Build GRCh38
UniGene ID:
Hs.506216
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16144
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220473.2
NP_001207402.1
12
1704
NM_001220474.2
NP_001207403.1
11
1650
NM_001220475.2
NP_001207404.1
12
1516
NM_001220476.2
NP_001207405.1
12
1516
NM_001220477.2
NP_001207406.1
10
1523
NM_001220478.2
NP_001207407.1
9
1448
NM_001220479.2
NP_001207408.1
3
265
NM_001220480.2
NP_001207409.1
3
265
NM_001282536.1
NP_001269465.1
10
1211
NM_003625.4
NP_003616.2
13
1567
XM_006719670.3
XP_006719733.1
10
8759
XM_011538906.2
XP_011537208.1
11
1644
XM_011538907.2
XP_011537209.1
11
1644
XM_017020079.1
XP_016875568.1
13
1551
XM_017020080.1
XP_016875569.1
12
1698
XM_017020081.1
XP_016875570.1
12
1698
XM_017020082.1
XP_016875571.1
13
1551
XM_017020083.1
XP_016875572.1
12
1698
XM_017020084.1
XP_016875573.1
12
1698
XM_017020085.1
XP_016875574.1
12
1698
XM_017020086.1
XP_016875575.1
13
1551
XM_017020087.1
XP_016875576.1
12
1685
XM_017020088.1
XP_016875577.1
12
1931
XM_017020089.1
XP_016875578.1
11
1644
XM_017020090.1
XP_016875579.1
13
1551
XM_017020091.1
XP_016875580.1
12
1698
XM_017020092.1
XP_016875581.1
12
1698
XM_017020093.1
XP_016875582.1
12
1698
XM_017020094.1
XP_016875583.1
12
1476
XM_017020095.1
XP_016875584.1
12
1698
XM_017020096.1
XP_016875585.1
11
1623
XM_017020097.1
XP_016875586.1
13
1551
XM_017020098.1
XP_016875587.1
13
1551
XM_017020099.1
XP_016875588.1
11
1644
XM_017020100.1
XP_016875589.1
12
1698
XM_017020101.1
XP_016875590.1
13
1551
XM_017020102.1
XP_016875591.1
11
1610
XM_017020103.1
XP_016875592.1
12
1698
XM_017020104.1
XP_016875593.1
12
1698
XM_017020105.1
XP_016875594.1
11
1623
XM_017020106.1
XP_016875595.1
12
1698
XM_017020107.1
XP_016875596.1
13
1551
XM_017020108.1
XP_016875597.1
13
1551
XM_017020109.1
XP_016875598.1
12
1698
XM_017020110.1
XP_016875599.1
11
1856
XM_017020111.1
XP_016875600.1
12
1698
XM_017020112.1
XP_016875601.1
10
1523
XM_017020113.1
XP_016875602.1
10
1523
XM_017020114.1
XP_016875603.1
10
1523
XM_017020115.1
XP_016875604.1
10
1523
XM_017020116.1
XP_016875605.1
10
1523
XM_017020117.1
XP_016875606.1
9
1258
XM_017020118.1
XP_016875607.1
11
1498
XM_017020119.1
XP_016875608.1
10
1523
XM_017020120.1
XP_016875609.1
10
1211
GenBank mRNA
AB210009.1
-
12
1702
AF034799.1
-
13
1575
AK123372.1
-
3
200
AK126971.1
-
11
1610
AK294505.1
-
9
1258
AK296380.1
-
3
236
AK299853.1
-
10
1472
AK302658.1
-
10
1182
BC104912.1
-
12
1500
8143915
BC143485.1
-
12
1500
9051993
DA533482.1
-
4
322
DA687865.1
-
3
249
DC322478.1
-
3
265
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
neurotransmitter secretion
glutamate secretion
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0