Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC43A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198810.1 | NP_001185739.1 | 12, 13 | 1446 | ||
NM_003627.5 | NP_003618.1 | 12, 13 | 1617 | |||
XM_005274358.4 | XP_005274415.1 | 13, 14 | 1757 | |||
XM_011545320.2 | XP_011543622.1 | 12, 13 | 1666 | |||
XM_011545321.2 | XP_011543623.1 | 10, 11 | 1233 | |||
XM_011545322.1 | XP_011543624.1 | 9, 10 | 1082 | |||
XM_017018451.1 | XP_016873940.1 | 13, 14 | 1645 | |||
XM_017018452.1 | XP_016873941.1 | 11, 12 | 1586 | |||
XM_017018453.1 | XP_016873942.1 | 10, 11 | 1423 | |||
GenBank mRNA | AB103033.1 | - | 12, 13 | 1584 | ||
AF045584.1 | - | 12, 13 | 1404 | |||
AK027732.1 | - | 10, 11 | 1182 | |||
BC001639.2 | - | 12, 13 | 1446 | 2959372 |