Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zfp746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100852.1 | NP_001094322.1 | 2 | 328 | ||
XM_006236419.2 | XP_006236481.1 | 2 | 770 | |||
XM_008762885.1 | XP_008761107.1 | 1 | 2659 | |||
XM_008762886.1 | XP_008761108.1 | 1 | 2659 | |||
XM_008762887.1 | XP_008761109.1 | 2 | 762 | |||
XM_008762888.1 | XP_008761110.1 | 1 | 2659 |