Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004312.2 | NP_004303.2 | 10, 11 | 768 | ||
XM_017029518.1 | XP_016885007.1 | 8, 9 | 745 | |||
GenBank mRNA | AF033105.1 | - | 10, 11 | 751 | ||
BC012096.1 | - | 10, 11 | 798 | 4645867 | ||
CR456849.1 | - | 8, 9 | 700 | |||
EU883571.1 | - | 8, 9 | 700 | |||
U03626.1 | - | 8, 9 | 700 |