Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080579.1 | NP_542146.1 | 2 | 943 | ||
XM_006237520.2 | XP_006237582.1 | 5 | 3126 | |||
XM_006237521.2 | XP_006237583.1 | 5 | 1407 | |||
XM_006237522.2 | XP_006237584.1 | 3 | 952 | |||
XM_006237523.2 | XP_006237585.1 | 3 | 1199 | |||
XM_006237524.1 | XP_006237586.1 | 3 | 1109 | |||
XM_008763379.1 | XP_008761601.1 | 4 | 1859 | |||
GenBank mRNA | BC089971.1 | - | 2 | 943 | 7315466 |
Set Membership: |
![]() |