Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprc5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079890.1 | NP_001073359.1 | 2 | 847 | ||
GenBank mRNA | BC086372.1 | - | 1 | 222 | ||
BC128767.1 | - | 2 | 847 | 7124284 |
Set Membership: |
![]() |