Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DGKZ SIRNA Assays ›
Gene Symbol:
DGKZ
Gene Name:
diacylglycerol kinase zeta
Gene Aliases:
DAGK5, DAGK6, DGK-ZETA, hDGKzeta
Chromosome Location:
Chr.11: 46332905 - 46380554 on Build GRCh38
UniGene ID:
Hs.502461
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16203
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DGKZ
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105540.1
NP_001099010.1
26
2927
NM_001199266.1
NP_001186195.1
25
2492
NM_001199267.1
NP_001186196.1
25
2489
NM_001199268.1
NP_001186197.1
24
2423
NM_003646.3
NP_003637.2
25
2492
NM_201532.2
NP_963290.1
25
2657
NM_201533.3
NP_963291.2
25
2393
XM_005253181.3
XP_005253238.1
25
2660
XM_005253182.3
XP_005253239.1
25
2391
XM_006718354.1
XP_006718417.1
26
3003
XM_006718355.2
XP_006718418.1
25
2265
XM_011520422.1
XP_011518724.1
24
2322
XM_011520423.1
XP_011518725.1
25
2370
XM_017018455.1
XP_016873944.1
26
3087
XM_017018456.1
XP_016873945.1
26
3085
XM_017018457.1
XP_016873946.1
25
3019
GenBank mRNA
AB209635.1
-
22
3855
AK124594.1
-
25
2657
AK124693.1
-
1
589
AK126970.1
-
25
2373
AK225774.1
-
25
2435
AK293615.1
-
4
366
AK294888.1
-
24
2241
AK300577.1
-
25
2135
AK303322.1
-
24
2162
AK312398.1
-
25
2311
BC008018.1
-
7
577
BC041770.1
-
25
2316
4811463
BX648433.1
-
25
2354
U51477.1
-
25
2323
U94905.1
-
26
2927
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein kinase C-activating G-protein coupled receptor signaling pathway
phosphorylation
cell migration
platelet activation
intracellular signal transduction
diacylglycerol kinase activity
protein binding
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-n6vvh:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0