Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003647.2 | NP_003638.1 | 11 | 1673 | ||
XM_011525394.2 | XP_011523696.1 | 11 | 1651 | |||
XM_011525395.2 | XP_011523697.1 | 11 | 1658 | |||
XM_017025243.1 | XP_016880732.1 | 11 | 2040 | |||
XM_017025244.1 | XP_016880733.1 | 10 | 1582 | |||
GenBank mRNA | AB209365.1 | - | 10 | 1323 | ||
AK293039.1 | - | 11 | 1620 | |||
BC130629.1 | - | 10 | 1517 | 40146660 | ||
BC130631.1 | - | 10 | 1517 | 40146662 | ||
U49379.1 | - | 11 | 1580 |