Gene Symbol: Tmem68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107903.1 | NP_001101373.1 | 7 | 1082 | ||
XM_006237827.2 | XP_006237889.1 | 7 | 1111 | |||
XM_006237832.2 | XP_006237894.1 | 7 | 1112 | |||
XM_008763539.1 | XP_008761761.1 | 7 | 1117 |