Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014447.2 | NP_001014447.1 | 6 | 1149 | ||
NM_001014448.2 | NP_001014448.1 | 6 | 1412 | |||
NM_003652.3 | NP_003643.2 | 5 | 1116 | |||
GenBank mRNA | AK056317.1 | - | 6 | 1149 | ||
AK127844.1 | - | 3 | 2485 | |||
BC006393.2 | - | 6 | 1316 | |||
BC018913.2 | - | 6 | 1316 | |||
BC080539.1 | - | 6 | 1083 | 5020194 | ||
CV026874.1 | - | 4 | 564 | |||
U83411.1 | - | 5 | 981 |