Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763819.1 | XP_008762041.1 | 14 | 3140 | ||
XM_008763820.1 | XP_008762042.1 | 14 | 3140 | |||
XM_008763821.1 | XP_008762043.1 | 14 | 3140 | |||
XM_008763822.1 | XP_008762044.1 | 13 | 3131 | |||
XM_008763823.1 | XP_008762045.1 | 14 | 3139 | |||
XM_008763824.1 | XP_008762046.1 | 13 | 3127 | |||
XM_008763825.1 | XP_008762047.1 | 13 | 3122 | |||
XM_008763826.1 | XP_008762048.1 | 12 | 3112 | |||
XM_008763827.1 | XP_008762049.1 | 13 | 3112 | |||
XM_008763828.1 | XP_008762050.1 | 14 | 3139 | |||
XM_008763829.1 | XP_008762051.1 | 14 | 3139 |