Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atpaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107959.1 | NP_001101429.1 | 7 | 866 | ||
XM_006238656.2 | XP_006238718.1 | 7 | 1682 |