Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKAPK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003668.3 | NP_003659.2 | 13, 14 | 1831 | ||
NM_139078.2 | NP_620777.1 | 13, 14 | 1837 | |||
XM_017020125.1 | XP_016875614.1 | 13, 14 | 1323 | |||
XM_017020126.1 | XP_016875615.1 | 11, 12 | 1237 | |||
XM_017020127.1 | XP_016875616.1 | 12, 13 | 1662 | |||
XM_017020128.1 | XP_016875617.1 | 12, 13 | 1662 | |||
XM_017020129.1 | XP_016875618.1 | 12, 13 | 1499 | |||
XM_017020130.1 | XP_016875619.1 | 12, 13 | 1267 | |||
XM_017020131.1 | XP_016875620.1 | 12, 13 | 1656 | |||
XM_017020132.1 | XP_016875621.1 | 13, 14 | 1710 | |||
XM_017020133.1 | XP_016875622.1 | 12, 13 | 1482 | |||
XM_017020134.1 | XP_016875623.1 | 12, 13 | 1308 | |||
XM_017020135.1 | XP_016875624.1 | 11, 12 | 1391 | |||
XM_017020136.1 | XP_016875625.1 | 12, 13 | 1476 | |||
XM_017020137.1 | XP_016875626.1 | 11, 12 | 1385 | |||
XM_017020138.1 | XP_016875627.1 | 8, 9 | 869 | |||
XM_017020139.1 | XP_016875628.1 | 12, 13 | 1449 | |||
GenBank mRNA | AK122767.1 | - | 13, 14 | 2081 | ||
AL110301.1 | - | 8, 9 | 933 | |||
BC000833.1 | - | 12, 13 | 1280 | |||
BC041049.1 | - | 13, 14 | 1818 | 4909021 | ||
BC047284.1 | - | 13, 14 | 1499 | 4997201 |