Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKAPK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003668.3 | NP_003659.2 | 5 | 863 | ||
NM_139078.2 | NP_620777.1 | 5 | 863 | |||
XM_017020125.1 | XP_016875614.1 | 5 | 355 | |||
XM_017020127.1 | XP_016875616.1 | 4 | 688 | |||
XM_017020128.1 | XP_016875617.1 | 4 | 688 | |||
XM_017020129.1 | XP_016875618.1 | 4 | 525 | |||
XM_017020130.1 | XP_016875619.1 | 4 | 293 | |||
XM_017020131.1 | XP_016875620.1 | 4 | 688 | |||
XM_017020132.1 | XP_016875621.1 | 5 | 721 | |||
GenBank mRNA | AK122767.1 | - | 5 | 1113 | ||
BC000833.1 | - | 4 | 306 | |||
BC041049.1 | - | 5 | 850 | 4909021 | ||
BC047284.1 | - | 5 | 531 | 4997201 |