Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itsn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750126.3 | XP_003750174.1 | 28 | 4271 | ||
XM_006239862.2 | XP_006239924.1 | 29 | 4370 | |||
XM_006239863.2 | XP_006239925.1 | 28 | 4289 | |||
XM_008764569.1 | XP_008762791.1 | 30 | 3907 | |||
XM_233945.8 | XP_233945.6 | 27 | 4189 |