Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004444.1 | NP_001004444.1 | 1 | 1803 | ||
XM_006240241.2 | XP_006240303.1 | 2 | 2017 | |||
XM_006240243.2 | XP_006240305.1 | 2 | 2024 | |||
XM_008764769.1 | XP_008762991.1 | 2 | 2017 | |||
GenBank mRNA | AY693992.1 | - | 1 | 1803 |