Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003679.4 | NP_003670.2 | 15 | 1700 | ||
XM_005273337.1 | XP_005273394.1 | 15 | 1771 | |||
XM_005273338.3 | XP_005273395.1 | 16 | 2053 | |||
XM_017002649.1 | XP_016858138.1 | 16 | 2044 | |||
XM_017002650.1 | XP_016858139.1 | 13 | 1468 | |||
GenBank mRNA | AF056032.1 | - | 15 | 1435 | ||
AK291558.1 | - | 15 | 1456 | |||
BC005297.1 | - | 15 | 1444 | 3934714 | ||
Y13153.1 | - | 15 | 1441 |