Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240365.2 | XP_006240427.1 | 13, 14 | 1705 | ||
XM_008764884.1 | XP_008763106.1 | 13, 14 | 1705 | |||
XM_008764885.1 | XP_008763107.1 | 13, 14 | 1624 | |||
XM_008764886.1 | XP_008763108.1 | 14, 15 | 1860 | |||
XM_008764887.1 | XP_008763109.1 | 14, 15 | 1720 | |||
XM_008764888.1 | XP_008763110.1 | 14, 15 | 1773 |