Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108064.1 | NP_001101534.1 | 6 | 1741 | ||
XM_006241061.1 | XP_006241123.1 | 8 | 4793 | |||
XM_006241062.2 | XP_006241124.1 | 8 | 4704 | |||
XM_008765161.1 | XP_008763383.1 | 10 | 4943 | |||
XM_008765162.1 | XP_008763384.1 | 10 | 4920 | |||
XM_008765163.1 | XP_008763385.1 | 10 | 4478 | |||
XM_008765164.1 | XP_008763386.1 | 10 | 4470 | |||
XM_008765166.1 | XP_008763388.1 | 8 | 4749 | |||
XM_008765167.1 | XP_008763389.1 | 8 | 2765 | |||
XM_008765168.1 | XP_008763390.1 | 6 | 2204 |