Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Midn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191577.1 | NP_001178506.1 | 1 | 199 | ||
XM_006240911.2 | XP_006240973.1 | 2 | 1856 | |||
XM_006240912.2 | XP_006240974.1 | 2 | 1583 | |||
XM_006240913.2 | XP_006240975.1 | 2 | 1856 | |||
XM_006240914.2 | XP_006240976.1 | 2 | 1856 |