Search Thermo Fisher Scientific
Gene Symbol: Slc39a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008356.1 | NP_001008357.1 | 4 | 1079 | ||
GenBank mRNA | BC086411.1 | - | 4 | 1079 | 7111777 | |
FQ221542.1 | - | 4 | 1123 |