Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106332.1 | NP_001099802.1 | 5, 6 | 458 | ||
GenBank mRNA | BC166771.1 | - | 5, 6 | 441 | 7934088 |