Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Chst11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108079.1 | NP_001101549.1 | 3 | 857 | ||
XM_006241200.2 | XP_006241262.1 | 3 | 2265 | |||
XM_008765238.1 | XP_008763460.1 | 4 | 2287 |