Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003693.3 | NP_003684.2 | 8 | 1445 | ||
NM_145350.2 | NP_663325.1 | 8 | 1445 | |||
NR_028075.2 | 8 | 1375 | ||||
NR_102409.1 | 8 | 1482 | ||||
XM_017025256.1 | XP_016880745.1 | 4 | 383 | |||
GenBank mRNA | AB052947.1 | - | 8 | 1309 | ||
AB052948.1 | - | 8 | 1412 | |||
AB052949.1 | - | 7 | 1784 | |||
AB052950.1 | - | 6 | 1111 | |||
AK291810.1 | - | 8 | 1388 | |||
AK298737.1 | - | 5 | 527 | |||
AK308904.1 | - | 4 | 383 | |||
AK310174.1 | - | 8 | 1405 | |||
BC039735.1 | - | 8 | 1413 | 6041360 | ||
D63483.1 | - | 8 | 1355 | |||
D86864.1 | - | 8 | 1379 | |||
DC429327.1 | - | 5 | 527 |