Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286673.1 | NP_001273602.1 | 4 | 655 | ||
NM_001286674.1 | NP_001273603.1 | 2 | 422 | |||
NM_001286675.1 | NP_001273604.1 | 3 | 506 | |||
NM_003702.4 | NP_003693.2 | 4 | 611 | |||
NM_170587.3 | NP_733466.1 | 5 | 1000 | |||
NR_104579.1 | 3 | 462 | ||||
XM_011517622.2 | XP_011515924.1 | 4 | 919 | |||
GenBank mRNA | AF060877.2 | - | 4 | 478 | ||
AF074979.1 | - | 3 | 395 | |||
AF366054.1 | - | 4 | 563 | |||
AF366055.1 | - | 3 | 414 | |||
AF366056.1 | - | 2 | 330 | |||
AF366057.1 | - | 4 | 428 | |||
AF493940.1 | - | 3 | 395 | |||
AK094503.1 | - | 3 | 462 | |||
AY046538.1 | - | 5 | 1000 | |||
BC015614.2 | - | 4 | 448 | |||
BC063490.1 | - | 4 | 601 | 5240001 | ||
JQ308180.1 | - | 4 | 467 |