Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212523.1 | NP_997688.1 | 23, 24 | 2743 | ||
GenBank mRNA | AY535015.1 | - | 23, 24 | 2743 | ||
FQ212220.1 | - | 2783 |