Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044245.1 | NP_001037710.1 | 9 | 899 | ||
GenBank mRNA | DQ238622.1 | - | 9 | 899 | ||
DQ238623.1 | - | 8 | 899 | |||
DQ238624.1 | - | 8 | 899 |