Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf20l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271439.1 | NP_001258368.1 | 1027 | |||
XM_008765533.1 | XP_008763755.1 | 7 | 822 | |||
XM_008765534.1 | XP_008763756.1 | 6 | 749 | |||
XM_008765535.1 | XP_008763757.1 | 5 | 671 | |||
XM_008765536.1 | XP_008763758.1 | 7 | 823 | |||
XM_008765537.1 | XP_008763759.1 | 5 | 671 | |||
XM_008765538.1 | XP_008763760.1 | 4 | 450 | |||
GenBank mRNA | BC096896.1 | - | 7 | 823 |