Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf20l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271439.1 | NP_001258368.1 | 801 | |||
XM_008765533.1 | XP_008763755.1 | 6 | 596 | |||
XM_008765534.1 | XP_008763756.1 | 5 | 523 | |||
XM_008765536.1 | XP_008763758.1 | 6 | 597 | |||
XM_008765538.1 | XP_008763760.1 | 3 | 224 | |||
GenBank mRNA | BC096896.1 | - | 6 | 597 |