Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270943.1 | NP_001257872.1 | 2 | 664 | ||
NM_001270944.1 | NP_001257873.1 | 2 | 628 | |||
NM_003709.3 | NP_003700.1 | 2 | 1107 | |||
XM_005246926.3 | XP_005246983.1 | 3 | 893 | |||
XM_006712815.3 | XP_006712878.1 | 3 | 897 | |||
XM_011512072.2 | XP_011510374.1 | 3 | 1071 | |||
XM_011512073.2 | XP_011510375.1 | 4 | 1079 | |||
XM_011512074.2 | XP_011510376.1 | 3 | 674 | |||
XM_011512075.1 | XP_011510377.1 | 2 | 1080 | |||
XM_017005161.1 | XP_016860650.1 | 3 | 893 | |||
XM_017005162.1 | XP_016860651.1 | 2 | 643 | |||
XM_017005163.1 | XP_016860652.1 | 2 | 1080 | |||
GenBank mRNA | AB015132.1 | - | 2 | 982 | ||
AK027371.1 | - | 2 | 808 | |||
AK297312.1 | - | 2 | 615 | |||
AK314439.1 | - | 2 | 1038 | |||
BX537890.1 | - | 2 | 664 | |||
CR542112.1 | - | 2 | 659 |