Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108104.2 | NP_001101574.2 | 6 | 782 | ||
GenBank mRNA | BC161934.1 | - | 6 | 780 | 9022144 |