Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PLPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003713.4 | NP_003704.3 | 2 | 804 | ||
GenBank mRNA | AB000889.1 | - | 2 | 242 | ||
AF017786.1 | - | 3 | 610 | |||
AF043329.1 | - | 2 | 529 | |||
AF480883.1 | - | 3 | 616 | |||
AK312439.1 | - | 2 | 762 | |||
BC009196.2 | - | 2 | 607 | 3960223 | ||
BT019589.1 | - | 2 | 236 | |||
U79294.1 | - | 3 | 610 |