Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PLPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003713.4 | NP_003704.3 | 2 | 763 | ||
GenBank mRNA | AB000889.1 | - | 2 | 201 | ||
AF017786.1 | - | 3 | 569 | |||
AF043329.1 | - | 2 | 488 | |||
AF480883.1 | - | 3 | 575 | |||
AK312439.1 | - | 2 | 721 | |||
BC009196.2 | - | 2 | 566 | 3960223 | ||
BT019589.1 | - | 2 | 195 | |||
U79294.1 | - | 3 | 569 |