Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108120.1 | NP_001101590.1 | 3 | 302 | ||
XM_006257360.2 | XP_006257422.1 | 4 | 383 | |||
XM_006257361.2 | XP_006257423.1 | 4 | 326 | |||
XM_008773417.1 | XP_008771639.1 | 3 | 301 | |||
GenBank mRNA | BC166561.1 | - | 3 | 238 | 7315205 |