Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034000.1 | NP_001029172.1 | 6 | 820 | ||
XM_006242735.2 | XP_006242797.1 | 6 | 1122 | |||
XM_006242736.2 | XP_006242798.1 | 6 | 773 | |||
XM_008766053.1 | XP_008764275.1 | 5 | 1197 | |||
GenBank mRNA | BC101928.1 | - | 6 | 820 | 7939116 |