Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201538.1 | NP_001188467.1 | 5 | 432 | ||
NM_001201539.1 | NP_001188468.1 | 5 | 517 | |||
NM_004042.4 | NP_004033.2 | 5 | 398 | |||
XM_011545522.1 | XP_011543824.1 | 5 | 384 | |||
XM_011545523.1 | XP_011543825.1 | 5 | 384 | |||
XM_011545524.1 | XP_011543826.1 | 5 | 384 | |||
XM_017029527.1 | XP_016885016.1 | 5 | 398 | |||
XM_017029528.1 | XP_016885017.1 | 5 | 384 | |||
GenBank mRNA | BC022389.1 | - | 5 | 517 | 4609825 | |
BP274206.1 | - | 5 | 432 | |||
BP277542.1 | - | 5 | 430 | |||
BQ228278.1 | - | 4 | 263 | |||
DA156994.1 | - | 5 | 399 | |||
DA628671.1 | - | 5 | 431 | |||
X97868.1 | - | 6 | 369 |