Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dscaml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108141.1 | NP_001101611.1 | 2 | 503 |
Set Membership: |
G-protein coupled receptor |