Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201538.1 | NP_001188467.1 | 6 | 925 | ||
NM_001201539.1 | NP_001188468.1 | 6 | 1010 | |||
NM_004042.4 | NP_004033.2 | 6 | 891 | |||
XM_011545522.1 | XP_011543824.1 | 6 | 877 | |||
XM_011545523.1 | XP_011543825.1 | 6 | 877 | |||
XM_011545524.1 | XP_011543826.1 | 6 | 877 | |||
XM_017029527.1 | XP_016885016.1 | 6 | 891 | |||
XM_017029528.1 | XP_016885017.1 | 6 | 877 | |||
GenBank mRNA | BC022389.1 | - | 6 | 1010 | 4609825 | |
BQ228278.1 | - | 5 | 756 | |||
BX107362.1 | - | 1 | 357 | |||
X97868.1 | - | 7 | 862 |