Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766402.1 | XP_008764624.1 | 52 | 7027 | ||
XM_008766403.1 | XP_008764625.1 | 52 | 9065 | |||
XM_008766404.1 | XP_008764626.1 | 52 | 7027 | |||
XM_008766405.1 | XP_008764627.1 | 51 | 6907 | |||
GenBank mRNA | FQ222849.1 | - | 2 | 83 | ||
FQ228515.1 | - | 2 | 83 |