Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243882.2 | XP_006243944.1 | 4 | 1493 | ||
XM_006243883.2 | XP_006243945.1 | 4 | 1550 | |||
XM_006243884.2 | XP_006243946.1 | 4 | 1490 | |||
XM_006243885.2 | XP_006243947.1 | 4 | 1601 |
Gene Symbol: LOC103693173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766661.1 | XP_008764883.1 | 3 | 683 |