Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003743.4 | NP_003734.3 | 12 | 2791 | ||
NM_147223.2 | NP_671756.1 | 12 | 2791 | |||
NM_147233.2 | NP_671766.1 | 12 | 2791 | |||
XM_005264625.1 | XP_005264682.1 | 14 | 3067 | |||
XM_005264626.1 | XP_005264683.1 | 13 | 2821 | |||
XM_005264628.1 | XP_005264685.1 | 13 | 2821 | |||
XM_017005168.1 | XP_016860657.1 | 14 | 3067 | |||
XM_017005169.1 | XP_016860658.1 | 14 | 3067 | |||
GenBank mRNA | AJ000881.1 | - | 12 | 2734 | ||
AJ000882.1 | - | 12 | 2734 | |||
AK226063.1 | - | 12 | 2746 | |||
AK290019.1 | - | 13 | 2821 | |||
BC111533.1 | - | 11 | 2628 | 40027438 | ||
BC111534.1 | - | 11 | 2628 | 40027439 | ||
U40396.1 | - | 2 | 1449 | |||
U59302.1 | - | 12 | 2727 | |||
U90661.1 | - | 10 | 2575 |