Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005743.1 | NP_001005743.1 | 7, 8 | 633 | ||
NM_001005744.1 | NP_001005744.1 | 7, 8 | 633 | |||
NM_001005745.1 | NP_001005745.1 | 6, 7 | 600 | |||
NM_001320114.1 | NP_001307043.1 | 7, 8 | 653 | |||
NM_003744.5 | NP_003735.3 | 6, 7 | 600 | |||
GenBank mRNA | AF015040.1 | - | 7, 8 | 591 | ||
AF108092.1 | - | 5, 6 | 418 | |||
AF171938.1 | - | 7, 8 | 583 | |||
AF171939.1 | - | 7, 8 | 583 | |||
AF171940.1 | - | 6, 7 | 550 | |||
AF171941.1 | - | 6, 7 | 550 | |||
AK023670.1 | - | 1, 2 | 670 | |||
AK226069.1 | - | 1, 2 | 2169 | |||
AK304193.1 | - | 5, 6 | 436 | |||
AK315130.1 | - | 5, 6 | 481 | |||
BC020788.1 | - | 5, 6 | 481 | |||
BC068476.1 | - | 6, 7 | 600 | 4822300 | ||
BX248073.1 | - | 6, 7 | 490 | |||
BX248775.1 | - | 5, 6 | 454 | |||
BX248781.1 | - | 5, 6 | 495 | |||
BX647137.1 | - | 7, 8 | 592 | |||
EU265734.1 | - | 5, 6 | 352 | |||
EU265735.1 | - | 4, 5 | 319 | |||
EU265736.1 | - | 5, 6 | 352 | |||
EU265737.1 | - | 4, 5 | 319 |