Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244701.2 | XP_006244763.1 | 18 | 4218 | ||
XM_006244702.2 | XP_006244764.1 | 18 | 4218 | |||
XM_006244703.2 | XP_006244765.1 | 18 | 4218 | |||
XM_006244704.2 | XP_006244766.1 | 18 | 4218 | |||
XM_006244705.2 | XP_006244767.1 | 18 | 4218 | |||
XM_006244706.2 | XP_006244768.1 | 18 | 4219 | |||
XM_006244707.2 | XP_006244769.1 | 22 | 2837 | |||
XM_006244708.2 | XP_006244770.1 | 18 | 4219 | |||
XM_006244713.2 | XP_006244775.1 | 25 | 3490 | |||
XM_006244714.2 | XP_006244776.1 | 22 | 2791 | |||
XM_006244715.2 | XP_006244777.1 | 22 | 2791 | |||
XM_006244716.2 | XP_006244778.1 | 12 | 4992 | |||
XM_008766964.1 | XP_008765186.1 | 23 | 2873 | |||
XM_008766965.1 | XP_008765187.1 | 12 | 4977 | |||
XM_008766966.1 | XP_008765188.1 | 12 | 4973 | |||
XM_008766967.1 | XP_008765189.1 | 25 | 3485 | |||
XM_008766968.1 | XP_008765190.1 | 22 | 2791 |