Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Arid5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034934.1 | NP_001030106.1 | 5 | 484 | ||
XM_006244806.2 | XP_006244868.1 | 7 | 786 | |||
XM_006244807.2 | XP_006244869.1 | 7 | 783 | |||
GenBank mRNA | BC105777.1 | - | 5 | 484 | 7931985 |