Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: EIF3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF020833.1 | - | 4 | 201 | ||
AF094850.1 | - | 4 | 219 | |||
AK291320.1 | - | 4 | 220 | |||
AK293374.1 | - | 4 | 304 | |||
AK296377.1 | - | 2 | 850 | |||
AK309944.1 | - | 4 | 218 | |||
AK311624.1 | - | 4 | 353 | |||
AK315896.1 | - | 4 | 225 | |||
AK316040.1 | - | 3 | 349 | |||
AK316335.1 | - | 3 | 355 | |||
BC000733.2 | - | 4 | 220 | 3504640 | ||
BC008469.1 | - | 4 | 221 | 4279770 | ||
BF701069.1 | - | 4 | 238 | |||
BT006889.1 | - | 4 | 178 | |||
CR457134.1 | - | 4 | 178 | |||
DC356593.1 | - | 4 | 243 | |||
U96074.1 | - | 4 | 187 |