Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103694495 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773355.1 | XP_008771577.1 | 3 | 273 | ||
XM_008773356.1 | XP_008771578.1 | 3 | 273 |
Gene Symbol: Fndc3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191722.1 | NP_001178651.1 | 2338 |