Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EIF4G3 SIRNA Assays ›
Gene Symbol:
EIF4G3
Gene Name:
eukaryotic translation initiation factor 4 gamma 3
Gene Aliases:
eIF-4G 3, eIF4G 3, eIF4GII
Chromosome Location:
Chr.1: 20806292 - 21177320 on Build GRCh38
UniGene ID:
Hs.467084
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16520
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EIF4G3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198801.1
NP_001185730.1
34
5278
NM_001198802.1
NP_001185731.1
34
5188
NM_003760.4
NP_003751.2
31
4804
XM_011542356.2
XP_011540658.1
37
5878
XM_011542361.2
XP_011540663.1
36
5767
XM_011542370.1
XP_011540672.1
37
5881
XM_011542371.1
XP_011540673.1
35
5623
XM_011542374.1
XP_011540676.1
34
5590
XM_011542375.2
XP_011540677.1
34
7178
XM_011542381.2
XP_011540683.1
24
3264
XM_017002672.1
XP_016858161.1
38
7416
XM_017002673.1
XP_016858162.1
36
5860
XM_017002674.1
XP_016858163.1
36
5857
XM_017002675.1
XP_016858164.1
36
5848
XM_017002676.1
XP_016858165.1
35
5827
XM_017002677.1
XP_016858166.1
35
5704
XM_017002678.1
XP_016858167.1
36
5770
XM_017002679.1
XP_016858168.1
36
5767
XM_017002680.1
XP_016858169.1
33
4797
XM_017002681.1
XP_016858170.1
36
5764
XM_017002682.1
XP_016858171.1
35
5746
XM_017002683.1
XP_016858172.1
35
5746
XM_017002684.1
XP_016858173.1
36
7272
XM_017002685.1
XP_016858174.1
35
5737
XM_017002686.1
XP_016858175.1
35
5734
XM_017002687.1
XP_016858176.1
35
5734
XM_017002688.1
XP_016858177.1
32
4763
XM_017002690.1
XP_016858179.1
35
5731
XM_017002691.1
XP_016858180.1
34
5716
XM_017002692.1
XP_016858181.1
34
5713
XM_017002693.1
XP_016858182.1
33
5584
XM_017002694.1
XP_016858183.1
35
5656
XM_017002695.1
XP_016858184.1
35
5653
XM_017002696.1
XP_016858185.1
34
5635
XM_017002697.1
XP_016858186.1
34
5632
XM_017002698.1
XP_016858187.1
34
5623
XM_017002700.1
XP_016858189.1
33
5602
XM_017002701.1
XP_016858190.1
33
5599
XM_017002702.1
XP_016858191.1
33
5533
XM_017002703.1
XP_016858192.1
31
4694
XM_017002704.1
XP_016858193.1
35
5746
XM_017002705.1
XP_016858194.1
36
5041
XM_017002706.1
XP_016858195.1
35
4930
XM_017002707.1
XP_016858196.1
33
4873
XM_017002708.1
XP_016858197.1
32
4762
GenBank mRNA
AB209119.1
-
34
5147
AF012072.2
-
31
4804
AK294883.1
-
25
4051
AK302873.1
-
22
3045
BC094683.1
-
29
3811
6188273
BC136643.1
-
31
4727
9020633
Z34918.1
-
15
1890
Back To Top
More Information
Panther Classification:
Molecular Function -
translation factor
translation initiation factor
translational protein
Gene Ontology Categories:
Function(s)
Process(es)
translational initiation
regulation of translational initiation
viral process
RNA cap binding
translation initiation factor activity
translation factor activity, RNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0