Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Porcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173355.1 | NP_001166826.1 | 12 | 1238 | ||
XM_006256716.2 | XP_006256778.1 | 11 | 1262 | |||
XM_006256717.2 | XP_006256779.1 | 11 | 1259 | |||
GenBank mRNA | BC158642.1 | - | 8 | 776 | 8362532 |