Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175600.1 | NP_783190.1 | 1 | 615 | ||
XM_008758669.1 | XP_008756891.1 | 1 | 705 |
Gene Symbol: Taar8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175601.1 | NP_783191.1 | 1 | 615 | ||
XM_006227718.2 | XP_006227780.2 | 2 | 681 | |||
XM_006227719.2 | XP_006227781.1 | 2 | 1103 | |||
XM_008758615.1 | XP_008756837.1 | 3 | 1261 |
Set Membership: |
G-protein coupled receptor |